Assay Details
Gene Symbol: LOC5572553 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654020.2 | XP_001654070.2 | 5 - 6 | 7192 | 73 | |
XM_021850029.1 | XP_021705721.1 | 6 - 7 | 7132 | 73 | ||
XM_021850030.1 | XP_021705722.1 | 6 - 7 | 7111 | 73 | ||
XM_021850031.1 | XP_021705723.1 | 6 - 7 | 7108 | 73 |
