Assay Details
Gene Symbol: LOC5567055 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651377.2 | XP_001651427.1 | 3 - 4 | 791 | 79 | |
XM_001651378.2 | XP_001651428.1 | 2 - 3 | 621 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
