Assay Details
Gene Symbol: LOC5564770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649072.2 | XP_001649122.1 | 2 - 3 | 675 | 87 | |
XM_021852095.1 | XP_021707787.1 | 2 - 3 | 673 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
