Assay Details
Gene Symbol: LOC5564770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649071.2 | XP_001649121.1 | 3 - 4 | 870 | 99 | |
XM_001649072.2 | XP_001649122.1 | 3 - 4 | 866 | 99 | ||
XM_001649073.2 | XP_001649123.1 | 3 - 4 | 869 | 99 | ||
XM_021852095.1 | XP_021707787.1 | 3 - 4 | 864 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
