Assay Details
Gene Symbol: LOC5564770 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649071.2 | XP_001649121.1 | 4 - 5 | 1101 | 111 | |
XM_001649072.2 | XP_001649122.1 | 4 - 5 | 1097 | 111 | ||
XM_001649073.2 | XP_001649123.1 | 4 - 5 | 1100 | 111 | ||
XM_021852095.1 | XP_021707787.1 | 4 - 5 | 1095 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
