Assay Details
Gene Symbol: LOC5570972 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659384.2 | XP_001659434.2 | 2 - 3 | 536 | 84 | |
XM_021850422.1 | XP_021706114.1 | 2 - 3 | 492 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
