Assay Details
Gene Symbol: LOC5569035 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846162.1 | XP_021701854.1 | 3 - 4 | 858 | 78 | |
XM_021846163.1 | XP_021701855.1 | 3 - 4 | 819 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
