Assay Details
Gene Symbol: LOC5576380 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662546.3 | XP_001662596.2 | 4 - 5 | 597 | 67 | |
XM_021844028.1 | XP_021699720.1 | 4 - 5 | 639 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
