Assay Details
Gene Symbol: LOC5574358 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842321.1 | XP_021698013.1 | 10 - 11 | 2317 | 68 | |
XM_021842322.1 | XP_021698014.1 | 10 - 11 | 2353 | 68 | ||
XM_021842323.1 | XP_021698015.1 | 10 - 11 | 2265 | 68 |

More Information
Set Membership: |
![]() ![]() |
