Assay Details
Gene Symbol: LOC5578570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852595.1 | XP_021708287.1 | 2 - 3 | 1549 | 77 | |
XM_021852596.1 | XP_021708288.1 | 2 - 3 | 1417 | 77 |
