Assay Details
Gene Symbol: LOC5578570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663910.2 | XP_001663960.2 | 2 - 3 | 1646 | 82 | |
XM_021852595.1 | XP_021708287.1 | 3 - 4 | 1844 | 82 | ||
XM_021852596.1 | XP_021708288.1 | 3 - 4 | 1712 | 82 |
