Assay Details
Gene Symbol: LOC5578570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663910.2 | XP_001663960.2 | 3 - 4 | 1798 | 67 | |
XM_021852595.1 | XP_021708287.1 | 4 - 5 | 1996 | 67 | ||
XM_021852596.1 | XP_021708288.1 | 4 - 5 | 1864 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
