Assay Details
Gene Symbol: LOC5566381 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846699.1 | XP_021702391.1 | 1 - 2 | 1161 | 94 | |
XM_021846700.1 | XP_021702392.1 | 1 - 2 | 1161 | 94 |
