Assay Details
Gene Symbol: LOC5569929 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658777.2 | XP_001658827.1 | 2 - 3 | 145 | 93 | |
XM_021848845.1 | XP_021704537.1 | 1 - 2 | 138 | 93 | ||
XM_021848950.1 | XP_021704642.1 | 2 - 3 | 75 | 93 | ||
XM_021849021.1 | XP_021704713.1 | 2 - 3 | 92 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021849021.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
