Assay Details
Gene Symbol: LOC5578344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656864.2 | XP_001656914.2 | 5 - 6 | 1165 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5578344 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656864.2 | XP_001656914.2 | 5 - 6 | 1165 | 102 |
Set Membership: |
![]() ![]() ![]() ![]() |