Assay Details
Gene Symbol: LOC5566854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651229.2 | XP_001651279.2 | 1 - 2 | 152 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001651229.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
