Assay Details
Gene Symbol: LOC5570709 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659252.2 | XP_001659302.2 | 1 - 2 | 379 | 72 | |
XM_021845181.1 | XP_021700873.1 | 1 - 2 | 381 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
