Assay Details
Gene Symbol: LOC5575871 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655823.2 | XP_001655873.1 | 2 - 3 | 879 | 52 | |
XM_021849172.1 | XP_021704864.1 | 2 - 3 | 879 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
