Assay Details
Gene Symbol: LOC5567858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651845.2 | XP_001651895.1 | 2 - 3 | 1292 | 91 | |
XM_021849417.1 | XP_021705109.1 | 2 - 3 | 996 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001651845.2, XM_021849417.1 transcripts.
Set Membership: |
![]() ![]() |
