Assay Details
Gene Symbol: LOC5567858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651845.2 | XP_001651895.1 | 4 - 5 | 1719 | 73 | |
XM_021849417.1 | XP_021705109.1 | 4 - 5 | 1423 | 73 |
