Assay Details
Gene Symbol: LOC5567858 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651845.2 | XP_001651895.1 | 5 - 6 | 1853 | 67 | |
XM_021849417.1 | XP_021705109.1 | 5 - 6 | 1557 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
