Assay Details
Gene Symbol: LOC5573254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654414.2 | XP_001654464.1 | 4 - 5 | 891 | 80 | |
XM_021847230.1 | XP_021702922.1 | 4 - 5 | 861 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
