Assay Details
Gene Symbol: LOC5573254 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654414.2 | XP_001654464.1 | 2 - 3 | 459 | 68 | |
XM_021847230.1 | XP_021702922.1 | 2 - 3 | 429 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
