Assay Details
Gene Symbol: LOC5570306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854285.1 | XP_021709977.1 | 15 - 16 | 4669 | 102 | |
XM_021854286.1 | XP_021709978.1 | 14 - 15 | 4595 | 102 | ||
XM_021854287.1 | XP_021709979.1 | 14 - 15 | 4726 | 102 | ||
XM_021854288.1 | XP_021709980.1 | 14 - 15 | 4535 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() |
