Assay Details
Gene Symbol: LOC5570306 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854285.1 | XP_021709977.1 | 12 - 13 | 4059 | 56 | |
XM_021854286.1 | XP_021709978.1 | 11 - 12 | 3985 | 56 | ||
XM_021854287.1 | XP_021709979.1 | 11 - 12 | 4116 | 56 | ||
XM_021854288.1 | XP_021709980.1 | 11 - 12 | 3925 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
