Assay Details
Gene Symbol: LOC5578022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848872.1 | XP_021704564.1 | 2 - 3 | 787 | 86 | |
XM_021848873.1 | XP_021704565.1 | 2 - 3 | 646 | 86 | ||
XM_021848874.1 | XP_021704566.1 | 2 - 3 | 659 | 86 | ||
XM_021848875.1 | XP_021704567.1 | 2 - 3 | 600 | 86 | ||
XM_021848876.1 | XP_021704568.1 | 2 - 3 | 420 | 86 | ||
XM_021848878.1 | XP_021704570.1 | 1 - 2 | 983 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
