Assay Details
Gene Symbol: LOC5568499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652276.2 | XP_001652326.1 | 1 - 2 | 412 | 65 | |
XM_001652277.2 | XP_001652327.1 | 1 - 2 | 411 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
