Assay Details
Gene Symbol: LOC5577595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656540.2 | XP_001656590.2 | 2 - 3 | 279 | 69 | |
XM_021846543.1 | XP_021702235.1 | 2 - 3 | 279 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
