Assay Details
Gene Symbol: LOC5577595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656540.2 | XP_001656590.2 | 1 - 2 | 162 | 72 | |
XM_021846543.1 | XP_021702235.1 | 1 - 2 | 162 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001656540.2, XM_021846543.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
