Assay Details
Gene Symbol: LOC5566597 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850317.1 | XP_021706009.1 | 2 - 3 | 520 | 85 | |
XM_021850318.1 | XP_021706010.1 | 2 - 3 | 289 | 85 | ||
XM_021850319.1 | XP_021706011.1 | 2 - 3 | 266 | 85 | ||
XM_021850320.1 | XP_021706012.1 | 2 - 3 | 520 | 85 |
