Assay Details
Gene Symbol: LOC5569429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852981.1 | XP_021708673.1 | 2 - 3 | 586 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5569429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852981.1 | XP_021708673.1 | 2 - 3 | 586 | 61 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |