Assay Details
Gene Symbol: LOC5565598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649896.2 | XP_001649946.1 | 2 - 3 | 348 | 70 | |
XM_021838095.1 | XP_021693787.1 | 2 - 3 | 348 | 70 | ||
XM_021838096.1 | XP_021693788.1 | 2 - 3 | 305 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001649896.2, XM_021838095.1, XM_021838096.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
