Assay Details
Gene Symbol: LOC5565598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649896.2 | XP_001649946.1 | 4 - 5 | 1744 | 76 | |
XM_021838095.1 | XP_021693787.1 | 4 - 5 | 1744 | 76 | ||
XM_021838096.1 | XP_021693788.1 | 4 - 5 | 1701 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
