Assay Details
Gene Symbol: LOC5573937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654923.2 | XP_001654973.1 | 2 - 3 | 483 | 78 | |
GenBank mRNA | AY431653.1 | - | 2 - 3 | 453 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
