Assay Details
Gene Symbol: LOC5570558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653169.2 | XP_001653219.1 | 3 - 4 | 1087 | 59 | |
XM_021849247.1 | XP_021704939.1 | 3 - 4 | 785 | 59 | ||
GenBank mRNA | AY431691.1 | - | 2 - 3 | 277 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
