Assay Details
Gene Symbol: LOC5576075 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662362.2 | XP_001662412.1 | 1 - 2 | 282 | 83 | |
GenBank mRNA | AY431765.1 | - | 1 - 2 | 243 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
