Assay Details
Gene Symbol: LOC5566140 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650457.2 | XP_001650507.1 | 2 - 3 | 401 | 63 | |
GenBank mRNA | AY431876.1 | - | 2 - 3 | 342 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
