Assay Details
Gene Symbol: LOC5580153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661588.3 | XP_001661638.1 | 6 | 2001 | 93 | |
GenBank mRNA | AY431904.1 | - | 1 | 522 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
