Assay Details
Gene Symbol: LOC5563691 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001647913.2 | XP_001647963.1 | 1 - 2 | 312 | 78 | |
GenBank mRNA | AY431905.1 | - | 1 - 2 | 231 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
