Assay Details
Gene Symbol: LOC5571150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653416.2 | XP_001653466.1 | 1 - 2 | 167 | 81 | |
GenBank mRNA | AY431967.1 | - | 1 - 2 | 77 | 81 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001653416.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
