Assay Details
Gene Symbol: LOC5570819 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846500.1 | XP_021702192.1 | 11 - 12 | 4360 | 92 | |
XM_021846501.1 | XP_021702193.1 | 11 - 12 | 4321 | 92 | ||
XM_021846502.1 | XP_021702194.1 | 11 - 12 | 4321 | 92 | ||
XM_021846503.1 | XP_021702195.1 | 11 - 12 | 4306 | 92 | ||
XM_021846504.1 | XP_021702196.1 | 11 - 12 | 4303 | 92 | ||
XM_021846505.1 | XP_021702197.1 | 11 - 12 | 4300 | 92 | ||
XM_021846506.1 | XP_021702198.1 | 11 - 12 | 4297 | 92 | ||
XM_021846507.1 | XP_021702199.1 | 11 - 12 | 4294 | 92 | ||
XM_021846508.1 | XP_021702200.1 | 11 - 12 | 4291 | 92 | ||
XM_021846509.1 | XP_021702201.1 | 11 - 12 | 4279 | 92 | ||
XM_021846511.1 | XP_021702203.1 | 11 - 12 | 4261 | 92 | ||
XM_021846512.1 | XP_021702204.1 | 11 - 12 | 4225 | 92 | ||
GenBank mRNA | AY431984.1 | - | 1 - 2 | 1084 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
