Assay Details
Gene Symbol: LOC5570819 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846500.1 | XP_021702192.1 | 12 - 13 | 4505 | 93 | |
XM_021846501.1 | XP_021702193.1 | 12 - 13 | 4466 | 93 | ||
XM_021846502.1 | XP_021702194.1 | 12 - 13 | 4466 | 93 | ||
XM_021846503.1 | XP_021702195.1 | 12 - 13 | 4451 | 93 | ||
XM_021846504.1 | XP_021702196.1 | 12 - 13 | 4448 | 93 | ||
XM_021846505.1 | XP_021702197.1 | 12 - 13 | 4445 | 93 | ||
XM_021846506.1 | XP_021702198.1 | 12 - 13 | 4442 | 93 | ||
XM_021846507.1 | XP_021702199.1 | 12 - 13 | 4439 | 93 | ||
XM_021846508.1 | XP_021702200.1 | 12 - 13 | 4436 | 93 | ||
XM_021846509.1 | XP_021702201.1 | 12 - 13 | 4424 | 93 | ||
XM_021846511.1 | XP_021702203.1 | 12 - 13 | 4406 | 93 | ||
XM_021846512.1 | XP_021702204.1 | 12 - 13 | 4370 | 93 | ||
GenBank mRNA | AY431984.1 | - | 2 - 3 | 1229 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
