Assay Details
Gene Symbol: LOC5575808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662184.2 | XP_001662234.1 | 2 - 3 | 563 | 86 | |
XM_021852504.1 | XP_021708196.1 | 2 - 3 | 651 | 86 | ||
XM_021852505.1 | XP_021708197.1 | 2 - 3 | 664 | 86 | ||
XM_021852506.1 | XP_021708198.1 | 2 - 3 | 701 | 86 | ||
XM_021852507.1 | XP_021708199.1 | 2 - 3 | 665 | 86 | ||
XM_021852508.1 | XP_021708200.1 | 2 - 3 | 562 | 86 | ||
XM_021852509.1 | XP_021708201.1 | 2 - 3 | 561 | 86 | ||
GenBank mRNA | AY432050.1 | - | 1 - 2 | 320 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
