Assay Details
Gene Symbol: LOC5575808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662184.2 | XP_001662234.1 | 3 - 4 | 696 | 99 | |
XM_021852504.1 | XP_021708196.1 | 3 - 4 | 784 | 99 | ||
XM_021852505.1 | XP_021708197.1 | 3 - 4 | 797 | 99 | ||
XM_021852506.1 | XP_021708198.1 | 3 - 4 | 834 | 99 | ||
XM_021852507.1 | XP_021708199.1 | 3 - 4 | 798 | 99 | ||
XM_021852508.1 | XP_021708200.1 | 3 - 4 | 695 | 99 | ||
XM_021852509.1 | XP_021708201.1 | 3 - 4 | 694 | 99 | ||
GenBank mRNA | AY432050.1 | - | 2 - 3 | 453 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
