Assay Details
Gene Symbol: LOC5577779 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663494.2 | XP_001663544.1 | 1 - 2 | 143 | 69 | |
XM_001663495.2 | XP_001663545.1 | 1 - 2 | 143 | 69 | ||
XM_021854632.1 | XP_021710324.1 | 1 - 2 | 143 | 69 | ||
XM_021854633.1 | XP_021710325.1 | 1 - 2 | 143 | 69 | ||
XM_021854634.1 | XP_021710326.1 | 1 - 2 | 143 | 69 | ||
XM_021854635.1 | XP_021710327.1 | 1 - 2 | 143 | 69 | ||
XM_021854636.1 | XP_021710328.1 | 1 - 2 | 143 | 69 | ||
XM_021854637.1 | XP_021710329.1 | 1 - 2 | 61 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001663494.2, XM_001663495.2, XM_021854632.1, XM_021854633.1, XM_021854634.1, XM_021854635.1, XM_021854636.1, XM_021854637.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
