Assay Details
Gene Symbol: LOC5569506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658541.2 | XP_001658591.1 | 5 - 6 | 4529 | 64 | |
XM_021844791.1 | XP_021700483.1 | 5 - 6 | 4000 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
