Assay Details
Gene Symbol: LOC5571574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845637.1 | XP_021701329.1 | 12 - 13 | 5928 | 79 | |
XM_021845641.1 | XP_021701333.1 | 12 - 13 | 6225 | 79 | ||
XM_021845647.1 | XP_021701339.1 | 12 - 13 | 5925 | 79 | ||
XM_021845651.1 | XP_021701343.1 | 12 - 13 | 6186 | 79 | ||
XM_021845661.1 | XP_021701353.1 | 12 - 13 | 5889 | 79 | ||
XM_021845670.1 | XP_021701362.1 | 12 - 13 | 5598 | 79 | ||
XM_021845681.1 | XP_021701373.1 | 12 - 13 | 5562 | 79 | ||
XM_021845688.1 | XP_021701380.1 | 12 - 13 | 5964 | 79 | ||
XM_021845692.1 | XP_021701384.1 | 11 - 12 | 5952 | 79 | ||
XM_021845695.1 | XP_021701387.1 | 11 - 12 | 5919 | 79 | ||
XM_021845698.1 | XP_021701390.1 | 10 - 11 | 5907 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
