Assay Details
Gene Symbol: LOC5571574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845637.1 | XP_021701329.1 | 6 - 7 | 2810 | 90 | |
XM_021845641.1 | XP_021701333.1 | 6 - 7 | 3107 | 90 | ||
XM_021845647.1 | XP_021701339.1 | 6 - 7 | 2807 | 90 | ||
XM_021845651.1 | XP_021701343.1 | 6 - 7 | 3068 | 90 | ||
XM_021845661.1 | XP_021701353.1 | 6 - 7 | 2771 | 90 | ||
XM_021845670.1 | XP_021701362.1 | 6 - 7 | 2480 | 90 | ||
XM_021845681.1 | XP_021701373.1 | 6 - 7 | 2444 | 90 | ||
XM_021845688.1 | XP_021701380.1 | 6 - 7 | 2846 | 90 | ||
XM_021845692.1 | XP_021701384.1 | 5 - 6 | 2834 | 90 | ||
XM_021845695.1 | XP_021701387.1 | 6 - 7 | 2846 | 90 | ||
XM_021845698.1 | XP_021701390.1 | 5 - 6 | 2834 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
