Assay Details
Gene Symbol: LOC5571574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845695.1 | XP_021701387.1 | 7 - 8 | 4338 | 89 | |
XM_021845698.1 | XP_021701390.1 | 6 - 7 | 4326 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
