Assay Details
Gene Symbol: LOC5571574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845637.1 | XP_021701329.1 | 10 - 11 | 5288 | 64 | |
XM_021845641.1 | XP_021701333.1 | 10 - 11 | 5585 | 64 | ||
XM_021845647.1 | XP_021701339.1 | 10 - 11 | 5285 | 64 | ||
XM_021845651.1 | XP_021701343.1 | 10 - 11 | 5546 | 64 | ||
XM_021845661.1 | XP_021701353.1 | 10 - 11 | 5249 | 64 | ||
XM_021845670.1 | XP_021701362.1 | 10 - 11 | 4958 | 64 | ||
XM_021845681.1 | XP_021701373.1 | 10 - 11 | 4922 | 64 | ||
XM_021845688.1 | XP_021701380.1 | 10 - 11 | 5324 | 64 | ||
XM_021845692.1 | XP_021701384.1 | 9 - 10 | 5312 | 64 | ||
XM_021845695.1 | XP_021701387.1 | 9 - 10 | 5279 | 64 | ||
XM_021845698.1 | XP_021701390.1 | 8 - 9 | 5267 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
